accessibility menu, dialog, popup

UserName

CAS RN 99-94-5

IUPAC Name: 4-methylbenzoic acid
Synonyms: p-toluic acid cadmium salt 4-toluic acid zinc salt 4-toluic acid crithminic acid calcium salt 4-toluic acid barium salt 4-toluic acid potassium salt 4-toluic acid toluenecarboxylic acid
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: LPNBBFKOUUSUDB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(O)=O
Boiling Point 
  • (3)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)