accessibility menu, dialog, popup

UserName

CAS RN 99-75-2

IUPAC Name: methyl 4-methylbenzoate
Synonyms: methyl P-toluate
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: QSSJZLPUHJDYKF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C=C1
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)