missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluate, 99%

Catalog No. AC414961000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414961000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC414961000 Supplier Thermo Scientific Chemicals Supplier No. 414961000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-75-2 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-75-2
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 32.0°C to 35.0°C
Density 1.0600g/mL
Boiling Point 220.0°C to 223.0°C
Flash Point 90°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H10O2
Linear Formula CH3C6H4COOCH3
Quantity 100 g
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in ethanol,ether
Molecular Weight (g/mol) 150.18
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl p-toluate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-784-1
RTECSNumber XU2150000
TSCA TSCA

RUO – Research Use Only