missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-Toluate 98.0+%, TCI America™

Catalog No. T0297500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0297500G 500 g
T029725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0297500G Supplier TCI America Supplier No. T0297500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-75-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC

Specifications

Color White
Boiling Point 223°C
Quantity 500 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Methyl p-Toluate
EINECSNumber (3)-1335&(3)-1380
RTECSNumber XU2150000
TSCA Yes