missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluate, 99%

Catalog No. AAB2311136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2311136 500 g
AAB2311122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2311136 Supplier Thermo Scientific Chemicals Supplier No. B2311136

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-75-2 | C9H10O2 | 150.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-75-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00008441
InChI Key QSSJZLPUHJDYKF-UHFFFAOYSA-N
Synonym methyl p-toluate, methyl-p-toluate, p-carbomethoxytoluene, methyl 4-toluate, 4-methylbenzoic acid methyl ester, methyl p-methylbenzoate, benzoic acid, 4-methyl-, methyl ester, 4-methoxycarbonyl toluene, p-toluic acid, methyl ester, methyl p-toluenecarboxylate
PubChem CID 7455
IUPAC Name methyl 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 32°C to 35°C
Density 1.06
Boiling Point 221°C to 224°C
Flash Point 89°C (192°F)
Odor Stench
Quantity 500 g
Beilstein 1100609
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material Methyl p-toluate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-784-1
RTECSNumber XU2150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only