CAS RN 99-35-4
IUPAC Name:
1,3,5-trinitrobenzene
Synonyms:
trinitrobenzene s-trinitrobenzene TNB sym-trinitrobenzene
Molecular Weight (g/mol):
213.11
Molecular Formula:
C6H3N3O6
InChi Key:
UATJOMSPNYCXIX-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
1
–
1
of
1
results
1
1,3,5-Trinitrobenzene (wetted with ca. 40% Water) (unit weight on dry weight basis) 98.0+%, TCI America™
CAS: 99-35-4 Molecular Formula: C6H3N3O6 Molecular Weight (g/mol): 213.105 MDL Number: MFCD00059179 InChI Key: UATJOMSPNYCXIX-UHFFFAOYSA-N Synonym: trinitrobenzene,sym-trinitrobenzene,s-trinitrobenzene,benzite,syn-trinitrobenzene,trinitrobenzeen,trinitrobenzol,benzene, 1,3,5-trinitro,benzene, trinitro-wet,rcra waste number u234 PubChem CID: 7434 ChEBI: CHEBI:48113 IUPAC Name: 1,3,5-trinitrobenzene SMILES: C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])[N+](=O)[O-]