accessibility menu, dialog, popup

UserName

CAS RN 98800-10-3

IUPAC Name: methyl thieno[3,2-b]thiophene-2-carboxylate
Synonyms: methyl thieno[3,2-b]thiophene-2-carboxylate
Molecular Weight (g/mol): 198.25
Molecular Formula: C8H6O2S2
InChi Key: VCLNJFSCOKHCEJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC2=C(S1)C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results