accessibility menu, dialog, popup

UserName

CAS RN 98474-78-3

CAS RN 98474-78-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)