missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Pseudoginsenoside RT5, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111001807
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001807 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001807 Supplier Merck Emd Millipore Supplier No. PHL8061910MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 98474-78-3
Product Line phyproof
Linear Formula C36H62O10
MDL Number MFCD10566641
Type Reference Substance
Quantity 10 mg
Formula Weight 654.87
Percent Purity ≥95% (HPLC)
Grade Primary Reference Standard

phyproof is a registered trademark of PhytoLab GmbH & Co. KG