CAS RN 98-66-8
CAS RN 98-66-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chlorobenzenesulfonic Acid Hydrate 98.0+%, TCI America™
CAS: 98-66-8 Molecular Formula: C6H5ClO3S Molecular Weight (g/mol): 192.613 MDL Number: MFCD00065342 InChI Key: RJWBTWIBUIGANW-UHFFFAOYSA-N Synonym: p-chlorobenzenesulfonic acid,4-chloro-benzenesulfonic acid,benzenesulfonic acid, 4-chloro,p-chlorophenylsulfonic acid,unii-5cn6b87ik2,4-chlorobenzenesulphonic acid,benzenesulfonic acid, p-chloro,4-chlorobenzene sulphonic acid,4-chlorobenzene sulfonic acid,closylate PubChem CID: 7400 IUPAC Name: 4-chlorobenzenesulfonic acid SMILES: C1=CC(=CC=C1S(=O)(=O)O)Cl