missing translation for 'onlineSavingsMsg'
Learn More

α-Terpineol, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019321
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019321 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019321 Supplier Merck Emd Millipore Supplier No. 04899250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity of Melaleuca alternifolia (tea tree) essential oil. The reaction rate constant of α-terpineol with OH radical and ozone was found to be (1.9 +/-0.5)x 10-10 cm3 molecule-1s-1 and (3.0 +/-0.2) x 10-16 cm3 molecule-1s-1, respectively. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 98-55-5
Density 0.93 g/mL (at 25°C)
Linear Formula C10H18 O
MDL Number MFCD00001557
Melting Point 31°C to 35°C
Boiling Point 217°C to 218°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 154.25
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less