accessibility menu, dialog, popup

UserName

CAS RN 98-53-3

IUPAC Name: 4-tert-butylcyclohexan-1-one
Synonyms: 4-tert-butylcyclohexanone
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: YKFKEYKJGVSEIX-UHFFFAOYSA-N
SMILES: CC(C)(C)C1CCC(=O)CC1
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (2)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (6)