accessibility menu, dialog, popup

UserName

CAS RN 978-62-1

IUPAC Name: N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
Synonyms: N-[3,5-bis(trifluoromethyl)phenyl]-5-chloro-2-hydroxybenzamide
Molecular Weight (g/mol): 383.67
Molecular Formula: C15H8ClF6NO2
InChi Key: CHILCFMQWMQVAL-UHFFFAOYSA-N
SMILES: OC1=C(C=C(Cl)C=C1)C(=O)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results