accessibility menu, dialog, popup

UserName

SBr

CAS RN 97511-05-2

IUPAC Name: 6-bromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Synonyms: 6-bromo-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular Weight (g/mol): 263.15
Molecular Formula: C12H7BrS
InChi Key: GJXAVNQWIVUQOD-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1SC1=C2C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results