accessibility menu, dialog, popup

UserName

OHClClHO

CAS RN 97-23-4

IUPAC Name: 4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
Synonyms: trivex epa pesticide chemical code 055001 caswell no. 563 preventol GD dichlorphen sandocide dichloorfeen fungicide GM diphenthane 70 fungicide M
Molecular Weight (g/mol): 269.12
Molecular Formula: C13H10Cl2O2
InChi Key: MDNWOSOZYLHTCG-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1CC1=CC(Cl)=CC=C1O
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)