CAS RN 96886-55-4
CAS RN 96886-55-4
IUPAC Name:
2-amino-2-methylpent-4-enoic acid
Synonyms:
2-amino-2-methylpent-4-enoic acid
Molecular Weight (g/mol):
129.16
Molecular Formula:
C6H11NO2
InChi Key:
QMBTZYHBJFPEJB-UHFFFAOYNA-N
SMILES:
CC(N)(CC=C)C(O)=O
1
–
1
of
1
results
1
(S)-(-)-2-Amino-2-methyl-4-pentenoic acid, 98%, ee 99+%
CAS: 96886-55-4 Molecular Formula: C6H11NO2 Molecular Weight (g/mol): 129.159 MDL Number: MFCD00145248 InChI Key: QMBTZYHBJFPEJB-LURJTMIESA-N Synonym: s-2-amino-2-methylpent-4-enoic acid,s-2-amino-2-methyl-4-pentenoic acid,l-alpha-allylalanine,2s-2-amino-2-methylpent-4-enoic acid,alpha-allyl-l-ala,r-2-amino-2-methyl-4-pentenoic acid,alpha-methyl-l-allylglycine,s---alpha-allylalanine,4-pentenoic acid, 2-amino-2-methyl-, 2s,s---2-amino-2-methyl-4-pentenoic acid PubChem CID: 2724753 IUPAC Name: (2S)-2-amino-2-methylpent-4-enoic acid SMILES: CC(CC=C)(C(=O)O)N