accessibility menu, dialog, popup

UserName

H3C(S)(S)OO(S)H3COO(S)H3CCH3HNO

CAS RN 96829-58-2

IUPAC Name: (2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl (2S)-2-formamido-4-methylpentanoate
Synonyms: orlistat roche brand of orlistat xenical (-)-tetrahydrolipstatin orlipastatum tetrahydrolipstatin 奥利司他 orlistat (product) orlipastat orlistat preparation
Molecular Weight (g/mol): 495.75
Molecular Formula: C29H53NO5
InChi Key: AHLBNYSZXLDEJQ-FWEHEUNISA-N
SMILES: CCCCCCCCCCC[C@@H](C[C@@H]1OC(=O)[C@H]1CCCCCC)OC(=O)[C@H](CC(C)C)NC=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)