accessibility menu, dialog, popup

UserName

CAS RN 968-21-8

H3CH3C(S)H2NOHN(S)OHOHO

CAS RN 968-21-8

IUPAC Name: (2S)-2-[(2S)-2-amino-4-methylpentanamido]-3-(4-hydroxyphenyl)propanoic acid
Synonyms: (2S)-2-[(2S)-2-amino-4-methylpentanamido]-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 294.35
Molecular Formula: C15H22N2O4
InChi Key: LHSGPCFBGJHPCY-STQMWFEESA-N
SMILES: CC(C)C[C@H](N)C(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results