accessibility menu, dialog, popup

UserName

HOOHONOO

CAS RN 96-97-9

IUPAC Name: 2-hydroxy-5-nitrobenzoic acid
Synonyms: 5-nitrosalicylic acid anilotic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: PPDRLQLKHRZIJC-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1O)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)