missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrosalicylic Acid 98.0+%, TCI America™

Catalog No. N025725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N025725G 25 g
N0257250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N025725G Supplier TCI America Supplier No. N025725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96-97-9
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00007338
InChI Key PPDRLQLKHRZIJC-UHFFFAOYSA-N
Synonym 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
PubChem CID 7318
ChEBI CHEBI:61281
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O

Specifications

Melting Point 236°C
Color Yellow
Quantity 25 g
Formula Weight 183.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrosalicylic Acid