missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrosalicylic acid, 98%

Catalog No. AAA1301022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1301022 100 g
AAA1301006 5 g
AAA1301014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1301022 Supplier Thermo Scientific Chemicals Supplier No. A1301022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-97-9 | C7H5NO5 | 183.119 g/mol

5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrosalicylic acid is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as an important organic, pharmaceutical intermediate.

Solubility
Soluble in water 1g/1475ml .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong bases.

Chemical Identifiers

CAS 96-97-9
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00007338
InChI Key PPDRLQLKHRZIJC-UHFFFAOYSA-N
Synonym 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
PubChem CID 7318
ChEBI CHEBI:61281
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O

Specifications

Melting Point 228°C to 236°C
Density 1.65
Quantity 100 g
Beilstein 2213719
Merck Index 14,6632
Solubility Information Soluble in water 1g/1475ml .
Formula Weight 183.12
Percent Purity 98%
Chemical Name or Material 5-Nitrosalicylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-548-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only