accessibility menu, dialog, popup

UserName

Explore All Our Special Offers, All in One Place

Save Today

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 96-26-4

OHOHO

CAS RN 96-26-4

IUPAC Name: 1,3-dihydroxypropan-2-one
Synonyms: dihydroxyacetone vitadye ketochromin a,a'-dihydroxyacetone soleal aliphatic ketone summers brand of dihydroxyacetone Otan viticolor chromelin
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: RXKJFZQQPQGTFL-UHFFFAOYSA-N
SMILES: OCC(=O)CO
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.