accessibility menu, dialog, popup

UserName

H3COO(E)

CAS RN 959-33-1

IUPAC Name: (2E)-3-(4-methoxyphenyl)-1-phenylprop-2-en-1-one
Synonyms: methoxychalcone
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: XUFXKBJMCRJATM-FMIVXFBMSA-N
SMILES: COC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)