CAS RN 95809-78-2
CAS RN 95809-78-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6,8-Bis(benzylthio)octanoic Acid 98.0+%, TCI America™
CAS: 95809-78-2 Molecular Formula: C22H28O2S2 Molecular Weight (g/mol): 388.58 MDL Number: MFCD22420826 InChI Key: ZYRLHJIMTROTBO-UHFFFAOYNA-N Synonym: CPI 613 PubChem CID: 24770514 IUPAC Name: 6,8-bis(benzylsulfanyl)octanoic acid SMILES: OC(=O)CCCCC(CCSCC1=CC=CC=C1)SCC1=CC=CC=C1