accessibility menu, dialog, popup

UserName

CAS RN 95648-78-5

CH3NOHNO(R)(S)(R)OOPOOOPOOOHONaNaNa

CAS RN 95648-78-5

IUPAC Name: trisodium ({[(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl phosphonato}oxy)phosphonate
Synonyms: trisodium thymidine 5'-diphosphate
Molecular Weight (g/mol): 468.13
Molecular Formula: C10H13N2Na3O11P2
InChi Key: HBYPEEZBYHQLDM-SPSULGLQSA-K
SMILES: [Na+].[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])([O-])=O)O2)C(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results