missing translation for 'onlineSavingsMsg'
Learn More

Ranolazine Dihydrochloride 98.0+%, TCI America™

Catalog No. R01125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01125G 5 g
R01121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01125G Supplier TCI America Supplier No. R01125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95635-56-6
Molecular Formula C24H35Cl2N3O4
Molecular Weight (g/mol) 500.46
MDL Number MFCD03788770
InChI Key RJNSNFZXAZXOFX-UHFFFAOYNA-N
Synonym ranolazine dihydrochloride, ranolazine hcl, ranolazine hydrochloride, renolazine, n-2,6-dimethylphenyl-2-4-2-hydroxy-3-2-methoxyphenoxy propyl piperazin-1-yl acetamide dihydrochloride, ranolazine hydrochloride usan, ranolazine 2hcl, +--4-2-hydroxy-3-o-methoxyphenoxy propyl-1-piperazineaceto-2',6'-xylidide dihydrochloride
PubChem CID 71279
IUPAC Name dihydrogen N-(2,6-dimethylphenyl)-2-{4-[2-hydroxy-3-(2-methoxyphenoxy)propyl]piperazin-1-yl}acetamide dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC1=CC=CC=C1OCC(O)CN1CCN(CC(=O)NC2=C(C)C=CC=C2C)CC1

Specifications

Melting Point 227°C
Color White
Quantity 5 g
Formula Weight 500.46
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ranolazine Dihydrochloride
RTECSNumber TK7845370
TSCA No