accessibility menu, dialog, popup

UserName

BrBr

CAS RN 952604-26-1

IUPAC Name: 1-bromo-4-(4-bromophenyl)naphthalene
Synonyms: 1-bromo-4-(4-bromophenyl)naphthalene
Molecular Weight (g/mol): 362.06
Molecular Formula: C16H10Br2
InChi Key: APZXILAQCHOANP-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC=C(Br)C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results