missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(4-bromophenyl)naphthalene 97.0+%, TCI America™

Catalog No. B50565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50565G 5 g
B50561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50565G Supplier TCI America Supplier No. B50565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 952604-26-1
Molecular Formula C16H10Br2
Molecular Weight (g/mol) 362.064
InChI Key APZXILAQCHOANP-UHFFFAOYSA-N
PubChem CID 57816069
IUPAC Name 1-bromo-4-(4-bromophenyl)naphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2Br)C3=CC=C(C=C3)Br

Specifications

Melting Point 148°C
Quantity 5 g
Formula Weight 362.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Bromo-4-(4-bromophenyl)naphthalene