accessibility menu, dialog, popup

UserName

CAS RN 95061-46-4

IUPAC Name: 1,1,2-triphenylethane-1,2-diol
Synonyms: 1,1,2-triphenylethane-1,2-diol
Molecular Weight (g/mol): 290.36
Molecular Formula: C20H18O2
InChi Key: GWVWUZJOQHWMFB-UHFFFAOYNA-N
SMILES: OC(C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results