accessibility menu, dialog, popup

UserName

CAS RN 95-74-9

IUPAC Name: 3-chloro-4-methylaniline
Synonyms: 3-chloro-P-toluidine C7H8ClN starlicide sulfate (2:1) starlicide ccris 152 starlicide monoacetate starlicide hydrochloride CPTH
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: RQKFYFNZSHWXAW-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1Cl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)