accessibility menu, dialog, popup

UserName

CAS RN 95-65-8

IUPAC Name: 3,4-dimethylphenol
Synonyms: 3,4-dimethylphenol 1,3,4-xylenol ccris 723
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: YCOXTKKNXUZSKD-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C=C1C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)