accessibility menu, dialog, popup

UserName

CAS RN 95-23-8

NH2HNOHN

CAS RN 95-23-8

IUPAC Name: 5-amino-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 5-amino-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 149.15
Molecular Formula: C7H7N3O
InChi Key: BCXSVFBDMPSKPT-UHFFFAOYSA-N
SMILES: NC1=CC=C2NC(=O)NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results