accessibility menu, dialog, popup

UserName

SN

CAS RN 95-16-9

IUPAC Name: 1,3-benzothiazole
Synonyms: benzothiazole fema number 3256 benzosulfonazole vangard BT benzothiazole (substance)
Molecular Weight (g/mol): 135.18
Molecular Formula: C7H5NS
InChi Key: IOJUPLGTWVMSFF-UHFFFAOYSA-N
SMILES: S1C=NC2=CC=CC=C12
Boiling Point 
  • (3)
  • (7)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (7)
Physical Form 
  • (10)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (3)
  • (7)
  • (4)
  • (6)
  • (3)
  • (7)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)