accessibility menu, dialog, popup

UserName

CAS RN 947725-04-4

IUPAC Name: 5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
Synonyms: 5-tert-butyl-1,3-dimethyl-2-(trifluoro-λ⁴-sulfanyl)benzene
Molecular Weight (g/mol): 250.32
Molecular Formula: C12H17F3S
InChi Key: VRTQPEYVMHATOA-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1S(F)(F)F)C(C)(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)