accessibility menu, dialog, popup

UserName

CAS RN 947-91-1

O

CAS RN 947-91-1

IUPAC Name: 2,2-diphenylacetaldehyde
Synonyms: benzeneacetaldehyde, α-phenyl-
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: HLLGFGBLKOIZOM-UHFFFAOYSA-N
SMILES: O=CC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results