accessibility menu, dialog, popup

UserName

O

CAS RN 946-33-8

IUPAC Name: 2-benzylcyclohexan-1-one
Synonyms: 2-benzylcyclohexan-1-one
Molecular Weight (g/mol): 188.27
Molecular Formula: C13H16O
InChi Key: CUYLPYBYCYQGCE-UHFFFAOYNA-N
SMILES: O=C1CCCCC1CC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)