accessibility menu, dialog, popup

UserName

FBrO

CAS RN 94569-84-3

IUPAC Name: 2-bromo-5-fluorobenzaldehyde
Synonyms: 2-bromo-5-fluorobenzaldehyde
Molecular Weight (g/mol): 203.01
Molecular Formula: C7H4BrFO
InChi Key: CJUCIKJLMFVWIS-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)C(C=O)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)