accessibility menu, dialog, popup

UserName

CAS RN 943-37-3

IUPAC Name: ethyl 5-nitrofuran-2-carboxylate
Synonyms: ethyl 5-nitrofuran-2-carboxylate
Molecular Weight (g/mol): 185.14
Molecular Formula: C7H7NO5
InChi Key: JMNXLAQKIHVFIC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(O1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results