accessibility menu, dialog, popup

UserName

H3CNNOOOCH2CH3(E/Z)

CAS RN 942230-11-7

IUPAC Name: 11-{4-[2-(4-butylphenyl)diazen-1-yl]phenoxy}undecyl 2-methylprop-2-enoate
Synonyms: 11-{4-[2-(4-butylphenyl)diazen-1-yl]phenoxy}undecyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 492.7
Molecular Formula: C31H44N2O3
InChi Key: YKWVMKBZDQOJNN-UHFFFAOYSA-N
SMILES: CCCCC1=CC=C(C=C1)N=NC1=CC=C(OCCCCCCCCCCCOC(=O)C(C)=C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results