CAS RN 942183-80-4
IUPAC Name:
1-(2-oxo-2-{4-[4-(pyrimidin-2-yl)phenyl]piperazin-1-yl}ethyl)-N-[3-(pyridin-4-yl)-1H-indazol-5-yl]pyrrolidine-3-carboxamide
Synonyms:
1-(2-oxo-2-{4-[4-(pyrimidin-2-yl)phenyl]piperazin-1-yl}ethyl)-N-[3-(pyridin-4-yl)-1H-indazol-5-yl]pyrrolidine-3-carboxamide
Molecular Weight (g/mol):
587.69
Molecular Formula:
C33H33N9O2
InChi Key:
HDAJDNHIBCDLQF-UHFFFAOYNA-N
SMILES:
O=C(CN1CCC(C1)C(=O)NC1=CC=C2NN=C(C2=C1)C1=CC=NC=C1)N1CCN(CC1)C1=CC=C(C=C1)C1=NC=CC=N1
1
–
1
of
1
results
1
Thermo Scientific Chemicals SCH772984, 95%
CAS: 942183-80-4 Molecular Formula: C33H33N9O2