CAS RN 94159-31-6
IUPAC Name:
2-(4-methyl-1,3-thiazol-5-yl)ethyl butanoate
Synonyms:
2-(4-methyl-1,3-thiazol-5-yl)ethyl butanoate
Molecular Weight (g/mol):
213.3
Molecular Formula:
C10H15NO2S
InChi Key:
GDRZNYCKSKHESZ-UHFFFAOYSA-N
SMILES:
CCCC(=O)OCCC1=C(C)N=CS1
1
–
1
of
1
results
1
2-(4-Methyl-5-thiazolyl)ethyl Butyrate 98.0+%, TCI America™
CAS: 94159-31-6 Molecular Formula: C10H15NO2S Molecular Weight (g/mol): 213.30 MDL Number: MFCD09032913 InChI Key: GDRZNYCKSKHESZ-UHFFFAOYSA-N Synonym: Butyric Acid 2-(4-Methyl-5-thiazolyl)ethyl Ester, 5-(2-Butyryloxyethyl)-4-methylthiazole PubChem CID: 3023838 IUPAC Name: 2-(4-methyl-1,3-thiazol-5-yl)ethyl butanoate SMILES: CCCC(=O)OCCC1=C(C)N=CS1