accessibility menu, dialog, popup

UserName

CAS RN 94108-56-2

IUPAC Name: 4-(trifluoromethoxy)benzene-1-sulfonyl chloride
Synonyms: 4-(trifluoromethoxy)benzenesulfonyl chloride
Molecular Weight (g/mol): 260.61
Molecular Formula: C7H4ClF3O3S
InChi Key: UHCDBMIOLNKDHG-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)