accessibility menu, dialog, popup

UserName

CAS RN 94-74-6

CH3ClOHOO

CAS RN 94-74-6

IUPAC Name: 2-(4-chloro-2-methylphenoxy)acetic acid
Synonyms: zelan MCPA chwastox extra hedonal M 4-chloro-o-cresoxyacetic acid U 46 M-fluid hormotuho caswell no. 557C legumex DB vesakontuho mcpa
Molecular Weight (g/mol): 200.62
Molecular Formula: C9H9ClO3
InChi Key: WHKUVVPPKQRRBV-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1OCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results