accessibility menu, dialog, popup

UserName

CAS RN 94-46-2

H3CCH3OO

CAS RN 94-46-2

IUPAC Name: 3-methylbutyl benzoate
Synonyms: isoamylbenzoate
Molecular Weight (g/mol): 192.26
Molecular Formula: C12H16O2
InChi Key: MLLAPOCBLWUFAP-UHFFFAOYSA-N
SMILES: CC(C)CCOC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results