accessibility menu, dialog, popup

UserName

CH3ONH2NS

CAS RN 94-45-1

IUPAC Name: 6-ethoxy-1,3-benzothiazol-2-amine
Synonyms: 6-ethoxy-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 194.25
Molecular Formula: C9H10N2OS
InChi Key: KOYJWFGMEBETBU-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2N=C(N)SC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results