accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 94-08-6

IUPAC Name: ethyl 4-methylbenzoate
Synonyms: ethyl P-toluate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: NWPWRAWAUYIELB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results