missing translation for 'onlineSavingsMsg'
Learn More

Ethyl p-Toluate 99.0+%, TCI America™

Catalog No. T0295500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0295500G 500 g
T029525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0295500G Supplier TCI America Supplier No. T0295500G

Chemical Identifiers

CAS 94-08-6
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00009117
InChI Key NWPWRAWAUYIELB-UHFFFAOYSA-N
Synonym ethyl p-toluate, ethyl p-methylbenzoate, benzoic acid, 4-methyl-, ethyl ester, 4-methylbenzoic acid ethyl ester, unii-xuf0sq8l2j, p-toluic acid ethyl ester, xuf0sq8l2j, p-toluic acid, ethyl ester, 4-methyl-benzoic acid ethyl ester, ethyl4-methylbenzoate
PubChem CID 66743
IUPAC Name ethyl 4-methylbenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C

Specifications

Color Colorless
Boiling Point 232°C
Quantity 500 g
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl p-Toluate
EINECSNumber (3)-1380
TSCA No