CAS RN 939-48-0
CAS RN 939-48-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isopropyl Benzoate 99.0+%, TCI America™
CAS: 939-48-0 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00048289 InChI Key: FEXQDZTYJVXMOS-UHFFFAOYSA-N Synonym: Benzoic Acid Isopropyl Ester PubChem CID: 13654 IUPAC Name: propan-2-yl benzoate SMILES: CC(C)OC(=O)C1=CC=CC=C1