accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CNNOH3COCH3

CAS RN 936250-17-8

IUPAC Name: 2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Synonyms: 2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Molecular Weight (g/mol): 266.1
Molecular Formula: C12H19BN2O4
InChi Key: RKHZHTLJEUBARP-UHFFFAOYSA-N
SMILES: COC1=NC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)