accessibility menu, dialog, popup

UserName

CoOOOO

CAS RN 932-69-4

IUPAC Name: λ²-cobalt(2+) dibenzoate
Synonyms: λ²-cobalt(2+) dibenzoate
Molecular Weight (g/mol): 301.16
Molecular Formula: C14H10CoO4
InChi Key: GAYAMOAYBXKUII-UHFFFAOYSA-L
SMILES: [Co++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results